MilliporeSigma
  • l-Citrulline and l-arginine supplementation retards the progression of high-cholesterol-diet-induced atherosclerosis in rabbits.
l-Citrulline and l-arginine supplementation retards the progression of high-cholesterol-diet-induced atherosclerosis in rabbits.
Proceedings of the National Academy of Sciences of the USA
Hayashi T