MilliporeSigma
  • miR-210 expression is associated with methionine-induced
    differentiation of trout satellite cells
miR-210 expression is associated with methionine-induced<BR/> differentiation of trout satellite cells
The Journal of Experimental Biology (2017)
Mary Latimer