MilliporeSigma
Antiviral activity of ovotransferrin derived peptides.
Biochemical and biophysical research communications (2005-04-23)
Francesco Giansanti, M Teresa Massucci, M Federica Giardi, Fabrizio Nozza, Emy Pulsinelli, Claudio Nicolini, Dario Botti, Giovanni Antonini
ABSTRACT
Ovotransferrin and lactoferrin are iron-binding proteins with antiviral and antibacterial activities related to natural immunity, showing marked sequence and structural homologies. The antiviral activity of two hen ovotransferrin fragments DQKDEYELL (hOtrf(219-227)) and KDLLFK (hOtrf(269-301) and hOtrf(633-638)) towards Marek's disease virus infection of chicken embryo fibroblasts is reported here. These fragments have sequence homology with two bovine lactoferrin fragments with antiviral activity towards herpes simplex virus, suggesting that these fragments could have a role for the exploitation of the antiviral activity of the intact proteins towards herpes viruses. NMR analysis showed that these peptides, chemically synthetized, did not possess any favourite conformation in solution, indicating that both the aminoacid sequence and the conformation they display in the intact protein are essential for the antiviral activity.
Sigma-Aldrich
Conalbumin from chicken egg white, suitable for cell culture, BioReagent