MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Tocopherol and selenite modulate the transplacental effects induced by sodium arsenite in hamsters.
Tocopherol and selenite modulate the transplacental effects induced by sodium arsenite in hamsters.
Reproductive toxicology (Elmsford, N.Y.) (2017-10-23)
Adriana Sampayo-Reyes, Reyes S Taméz-Guerra, Mario Bermúdez de León, Javier Vargas-Villarreal, Héctor Gerardo Lozano-Garza, Cristina Rodríguez-Padilla, Constanza Cortés, Ricard Marcos, Alba Hernández
ABSTRACT
Human studies suggest that in utero exposure to arsenic results in adverse pregnancy outcomes. The use of dietary supplements, such as sodium selenite (SS) or α-tocopherol succinate (α-TOS), is a reasonable approach to ameliorate such health effects. Sodium arsenite at 100ppm was administered via drinking water to female hamsters from gestational days 1 or 8 to the time of delivery. Viable fetuses, fetal resorptions and non-viable fetuses were recorded during and after pregnancy and total arsenic and its metabolites were characterized in pregnant animals, placentas and fetuses. Arsenic was found to accumulate in the placenta and fetus, increasing fetal mortality, non-viable fetuses and resorptions. Co-administration of SS and α-TOS significantly reduced the observed teratogenic effects. SS influenced arsenic biotransformation by reducing the MMA/InAs index and increasing the DMA/MMA, whereas α-TOS more likely exerts its protective effect through its potent antioxidant activity.
Sigma-Aldrich
D-α-Tocopherol succinate, semisynthetic, 1210 IU/g
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells