MilliporeSigma
  • Mesenchymal stem cells attenuate liver fibrosis by suppressing Th17 cells - an experimental study.
Mesenchymal stem cells attenuate liver fibrosis by suppressing Th17 cells - an experimental study.
Transplant international : official journal of the European Society for Organ Transplantation (2017-08-15)
Neda Milosavljevic, Marina Gazdic, Bojana Simovic Markovic, Aleksandar Arsenijevic, Jasmin Nurkovic, Zana Dolicanin, Nemanja Jovicic, Ilija Jeftic, Valentin Djonov, Nebojsa Arsenijevic, Miodrag L Lukic, Vladislav Volarevic
ABSTRACT
This study investigates molecular and cellular mechanisms involved in mesenchymal stem cell (MSC)-mediated modulation of IL-17 signaling during liver fibrosis. Mice received CCl
Sigma-Aldrich
1-Methyl-L-tryptophan, 95%