MilliporeSigma
  • A role for glutathione reductase and glutathione in the tolerance of Chlamydomonas reinhardtii to photo-oxidative stress.
A role for glutathione reductase and glutathione in the tolerance of Chlamydomonas reinhardtii to photo-oxidative stress.
Physiologia plantarum (2017-09-28)
Tsen-Hung Lin, Meng-Yuan Rao, Hao-Wen Lu, Chih-Wen Chiou, Shu-Tseng Lin, Hung-Wei Chao, Zhao-Liang Zheng, Hao-Chien Cheng, Tse-Min Lee
ABSTRACT
The role of glutathione reductase (GR; EC 1.6.4.2) in the tolerance of Chlamydomonas reinhardtii P.A. Dangeard to high-intensity light stress (HL, 1400 μmol m
Sigma-Aldrich
Protease Inhibitor Cocktail, for plant cell and tissue extracts, DMSO solution