MilliporeSigma
  • A rapid, naked-eye detection of hypochlorite and bisulfite using a robust and highly-photostable indicator dye Quinaldine Red in aqueous medium.
A rapid, naked-eye detection of hypochlorite and bisulfite using a robust and highly-photostable indicator dye Quinaldine Red in aqueous medium.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-10-19)
Tanoy Dutta, Falguni Chandra, Apurba L Koner
ABSTRACT
A "naked-eye" detection of health hazardous bisulfite (HSO
Sigma-Aldrich
Sodium hydrogen sulfate
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%