MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Purification of a Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferase and a Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase to homogeneity from rat liver.
Purification of a Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferase and a Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase to homogeneity from rat liver.
The Journal of biological chemistry (1982-11-25)
J Weinstein, U de Souza-e-Silva, J C Paulson
ABSTRACT
A Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferse and a Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase have been purified 23,000- and 860,000-fold to homogeneity from Triton CF-54 extracts of rat liver membranes. The two enzymes were concentrated by affinity chromatography on CDP-hexanolamine-agarose and resolved by NaCl gradient elution from the same adsorbent. Final purification of the Gal beta 1 to 4GlcNAc alpha 2 to 6 sialytransferase, the most abundant enzyme, was achieved by specific elution from CDP-agarose with CDP. The Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase was also purified further by CDP elution from CDP-agarose, but final purification required affinity chromatography on an adsorbent prepared by coupling asialoprothrombin to cyanogen bromide-activated agarose. Asialoprothrombin contains the terminal sequence Gal beta 1 to 3GlcNAc on N-linked oligosaccharides and is the best acceptor substrate of the enzyme (Km congruent to 6 microM). The Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase was found to bind to asialoprothrombin-agarose in the presence of CDP and could be eluted with a solution containing 0.2 M lactose and no CDP. Sodium dodecyl sulfate-gel electrophoresis of the Gal beta 1 to 4GlcNAc alpha 2 to 6 and Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferases revealed a single major protein band for each enzyme with apparent molecular weights of 40,500 and 44,000, respectively. Rabbit antibodies raised to the Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferase inhibit its enzymatic activity greater than 99% but caused little or no inhibition of Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialytransferase. Moreover, the Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferase quantitatively bound to a column containing antibody adsorbed to Protein A-agarose, while the Gal beta 1 to 3(4) GlcNAc alpha 2 to 3 sialyltransferase did not bind. This demonstrated that the two sialyltransferases are antigenically unrelated and formed the basis for removal of contaminating Gal beta 1 to 4GlcNAc alpha 2 to 6 sialyltransferase from solutions of the Gal beta 1 to 3(4)GlcNAc alpha 2 to 3 sialyltransferase. Enzymatic characterization of the two sialyltransferases suggests that their major biological roles are in the terminal glycosylation of N-linked oligosaccharides of glycoproteins. (Weinstein, J., de Souza-e-Silva, U., and Paulson J. C. (1982) J. Biol. Chem. 257, 13845-13853. The alpha 2 to 6 sialyltransferase efficiently forms the NeuAc alpha 2 to 6Gal beta 1 to 4GlcNAc sequence, and the alpha 2 to 3 sialyltransferase forms the NeuAc alpha 2 to 3Gal beta 1 to 3GlcNAc and NeuAc alpha 2 to 3Ga; beta 1 to 4GlcNAc sequences.
Sigma-Aldrich
Beta-galactoside alpha-2,6-sialyltransferase 1, ≥300 units/mg protein, ST6GAL1 human recombinant, expressed in HEK 293 cells