MilliporeSigma
  • Purification of camel liver catalase by zinc chelate affinity chromatography and pH gradient elution: An enzyme with interesting properties.
Purification of camel liver catalase by zinc chelate affinity chromatography and pH gradient elution: An enzyme with interesting properties.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-11-05)
Abdelbasset Chafik, Abdelkhalid Essamadi, Safinur Yildirim Çelik, Ahmet Mavi
ABSTRACT
Climate change and increasing temperatures are global concerns. Camel (Camelus dromedarius) lives most of its life under high environmental stress in the desert and represent ideal model for studying desert adaptation among mammals. Catalase plays a key role in protecting cells against oxidative stress. For the first time, catalase from camel liver was purified to homogeneity by zinc chelate affinity chromatography using pH gradient elution, a better separation was obtained. A purification fold of 201.81 with 1.17% yield and a high specific activity of 1132539.37U/mg were obtained. The native enzyme had a molecular weight of 268kDa and was composed of four subunits of equal size (65kDa). The enzyme showed optimal activity at a temperature of 45°C and pH 7.2. Thiol reagents, β-Mercaptoethanol and D,L-Dithiothreitol, inhibited the enzyme activity. The enzyme was inhibited by Al
Product Description
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 392 mPa.s (25 °C)