MilliporeSigma
  • Pre-operative trans-catheter arterial chemo-embolization increases hepatic artery thrombosis after liver transplantation - a retrospective study.
Pre-operative trans-catheter arterial chemo-embolization increases hepatic artery thrombosis after liver transplantation - a retrospective study.
Transplant international : official journal of the European Society for Organ Transplantation (2017-09-05)
Nicholas Gilbo, Laura Van Praet, Ina Jochmans, Mauricio Sainz-Barriga, Chris Verslype, Geert Maleux, Wim Laleman, Schalk van der Merwe, David Cassiman, Frederik Nevens, Diethard Monbaliu, Jacques Pirenne
ABSTRACT
Little is known about nonsurgical risk factors for hepatic artery thrombosis (HAT) after liver transplantation (LT). We determined risk factors for HAT occurring within 90 days post-LT and analysed the effect of HAT on graft and patient survival. Donor and recipient demographics, surgery-related data and outcome in transplants complicated by thrombosis (HAT+) and their matched controls (HAT-) were compared. Risk factors were assessed by univariate logistic regression. Median (IQR) is given. A total of 25 HAT occurred among 1035 adult LT (1/1997-12/2014) and 50 controls were manually matched. Donor and recipient demographics were similar. Pre-LT trans-catheter arterial chemo-embolization (TACE) was more frequent in HAT+ (HAT+ 20% vs. HAT- 4%, P = 0.037). HAT+ had longer implantation [HAT+ 88 min (76-108) vs. HAT- 77 min (66-93), P = 0.028] and surgery times [HAT+ 6.25 h (5.18-7.47) vs. HAT- 5.25 h (4.33-6.5), P = 0.001]. Early graft dysfunction and sepsis were more frequent in HAT+ and hospitalization longer. TACE had the greatest odds ratio in unadjusted analysis (OR: 6, 95% CI: 1.07-33.53, P = 0.03). All but seven grafts were lost after HAT (HAT+ 72% vs. HAT- 36%, P = 0.003); however, patient survival was unaffected (HAT+ 79.8% vs. HAT- 76%, P = 0.75). LT candidates undergoing TACE are at risk of developing HAT early after transplant.