MilliporeSigma
  • Epigallocatechin-3-gallate promotes apoptosis in human breast cancer T47D cells through down-regulation of PI3K/AKT and Telomerase.
Epigallocatechin-3-gallate promotes apoptosis in human breast cancer T47D cells through down-regulation of PI3K/AKT and Telomerase.
Pharmacological reports : PR (2017-06-25)
Maliheh Moradzadeh, Azar Hosseini, Saiedeh Erfanian, Hadi Rezaei
ABSTRACT
Green tea has antioxidant, anti-tumor and anti-bacterial properties. Epigallocatechin-3-gallate (EGCG) in green tea is highly active as a cancer chemopreventive agent. In this study, we designed a series of experiments to examine the effects of EGCG on proliferation and apoptosis of estrogen receptor α-positive breast cancer (T47D) cells. Cells were treated with EGCG (0-80μM) and tamoxifen (0-20μM), as the positive control, up to 72h. Cell viability was determined by MTT assay. Apoptosis investigated by real time PCR of apoptosis and survival (Bax, Bcl-2, p21, p53, PTEN, PI3K, AKT, caspase3 and caspase9 and hTERT) genes and by western blot of Bax/Bcl-2 proteins expressions. The results showed that EGCG decreased cell viability as concentration- and time-dependently. IC This study suggests that EGCG may be a useful adjuvant therapeutic agent for the treatment of breast cancer.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%