MilliporeSigma
  • Betamethasone for lung maturation: testing dose and formulation in fetal sheep.
Betamethasone for lung maturation: testing dose and formulation in fetal sheep.
American journal of obstetrics and gynecology (2007-11-06)
Alan H Jobe, Timothy J M Moss, Ilias Nitsos, Machiko Ikegami, Suhas G Kallapur, John P Newnham
ABSTRACT
We evaluated lung maturation responses to the mixture of betamethasone phosphate (Beta-PO4) + betamethasone acetate (Beta-Ac) in comparison to Beta-PO4 or Beta-Ac in fetal sheep. Ewes carrying singleton pregnancies at 122 days of gestation were randomized to single doses of 0.5 mg/kg Beta-PO4 + Beta-Ac, 0.25 mg/kg Beta-PO4, 0.5 mg/kg Beta-PO4, or 0.25 mg/kg Beta-Ac given 48 hours before delivery. These treatments were compared with saline placebo and 2 doses of 0.5 mg/kg Beta-PO4 + Beta-Ac that were given 48 and 24 hours before delivery. Fetal lung maturation was evaluated. The 2 doses of the Beta-PO4 + Beta-Ac mixture gave the best lung maturation. Single doses of the Beta-PO4 + Beta-Ac mixture and Beta-Ac alone also induced lung maturation. There were no consistent responses to either dose of Beta-PO4. Beta-PO4 alone is ineffective with the dosing schedule used; Beta-Ac can induce lung maturation. However, the best responses result from the mixture of Beta-PO4 + Beta-Ac.