MilliporeSigma
  • SUMOylated ORC2 Recruits a Histone Demethylase to Regulate Centromeric Histone Modification and Genomic Stability.
SUMOylated ORC2 Recruits a Histone Demethylase to Regulate Centromeric Histone Modification and Genomic Stability.
Cell reports (2016-04-08)
Chao Huang, Jinke Cheng, Tasneem Bawa-Khalfe, Xuebiao Yao, Y Eugene Chin, Edward T H Yeh
ABSTRACT
Origin recognition complex 2 (ORC2), a subunit of the ORC, is essential for DNA replication initiation in eukaryotic cells. In addition to a role in DNA replication initiation at the G1/S phase, ORC2 has been shown to localize to the centromere during the G2/M phase. Here, we show that ORC2 is modified by small ubiquitin-like modifier 2 (SUMO2), but not SUMO1, at the G2/M phase of the cell cycle. SUMO2-modification of ORC2 is important for the recruitment of KDM5A in order to convert H3K4me3 to H3K4me2, a "permissive" histone marker for α-satellite transcription at the centromere. Persistent expression of SUMO-less ORC2 led to reduced α-satellite transcription and impaired pericentric heterochromatin silencing, which resulted in re-replication of heterochromatin DNA. DNA re-replication eventually activated the DNA damage response, causing the bypass of mitosis and the formation of polyploid cells. Thus, ORC2 sustains genomic stability by recruiting KDM5A to maintain centromere histone methylation in order to prevent DNA re-replication.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)