MilliporeSigma
Identification of the
Biochemistry and biophysics reports (2016-07-20)
Takahiro Ide, Shota Mochiji, Noriko Ueki, Katsushi Yamaguchi, Shuji Shigenobu, Masafumi Hirono, Ken-Ichi Wakabayashi
ABSTRACT
The unicellular green alga
Sigma-Aldrich
Anti-Guinea Pig IgG (whole molecule)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution