MilliporeSigma
  • Palladium-catalyzed gamma-arylation of alpha,beta-unsaturated esters from silyl ketene acetals.
Palladium-catalyzed gamma-arylation of alpha,beta-unsaturated esters from silyl ketene acetals.
Angewandte Chemie (International ed. in English) (2010-07-08)
David S Huang, John F Hartwig