MilliporeSigma
  • Potential regulation of glioma through the induction of apoptosis signaling via Egl-9 family hypoxia-inducible factor 3.
Potential regulation of glioma through the induction of apoptosis signaling via Egl-9 family hypoxia-inducible factor 3.
Oncology letters (2017-03-31)
Ke Mao, Chao You, Ding Lei, Heng Zhang
ABSTRACT
Glioma is an aggressive form of brain cancer that occurs following the abnormal proliferation of glial cells. Although glioma cannot spread to other organs, the morbidity and mortality rates of the disease are high, even following surgery, radiotherapy and chemotherapy. The function of Egl-9 family hypoxia-inducible factor 3 (Egln3) in cancer is controversial, and it is debated as to whether Egln3 positively or negatively regulates tumors. In the present study, a mouse model of low-grade glioma was successfully established. Through the use of immunohistochemical and western blot analyses, it was demonstrated that Egln3 expression in glioma tissue performed an important role in regulation by amplifying the signals for apoptosis, as determined by an increase in DNA fragments. Furthermore, Egln3 expression was inhibited by the administration of dimethyloxalylglycine, and the downregulated expression of Egln3 had marked effects on the regulation of glioma through apoptosis. The present study therefore provides evidence of an association between Egln3 expression and apoptosis in low-grade glioma.
Sigma-Aldrich
Anti-BrdU Antibody, clone BU-1, culture supernatant, clone BU-1, Upstate®