MilliporeSigma
  • Tenotomy immobilization as a model to investigate skeletal muscle fibrosis (with emphasis on Secreted frizzled-related protein 2).
Tenotomy immobilization as a model to investigate skeletal muscle fibrosis (with emphasis on Secreted frizzled-related protein 2).
Physiological genomics (2016-04-27)
Uğur Akpulat, İlyas Onbaşılar, Y Çetin Kocaefe
ABSTRACT
The pathological endpoint of congenital and senile myopathies is chronic muscle degeneration characterized by the atrophy of contractile elements, accompanied by fibrosis and fatty infiltration of the interstitium. Tenotomy is the release of preload that causes abrupt shortening of the muscle and models atrophy and fibrosis without prominent inflammatory response. Fibrosis in the skeletal muscle is known to be triggered by transforming growth factor (TGF)-β, which is activated by inflammatory events. As these were lacking, tenotomy provided an opportunity to investigate transcriptional events on a background without inflammation. An unbiased look at the transcriptome of tenotomy-immobilized soleus muscle revealed that the majority of the transcriptional changes took place in the first 4 wk. Regarding atrophy, proteasomal and lysosomal pathways were actively involved in accompanying cathepsins and calpains in the breakdown of the macromolecular contractile machinery. The transcriptome provided clear-cut evidence for the upregulation of collagens and several extracellular matrix components that define fibrotic remodeling of the skeletal muscle architecture as well as activation of the fibro-adipogenic precursors. Concomitantly, Sfrp2, a Wnt antagonist as well as a procollagen processor, accompanied fibrosis in skeletal muscle with an expression that was stringently confined to the slow-twitch fibers. An interpreted mechanistic scenario construed the kinetic events initiated through the abnormal shortening of the muscle fibers as enough to trigger the resident latent TGF-β in the extracellular matrix, leading to the activation of fibroadipogenic precursors. As in the heart, Sfrp2 shows itself to be a therapeutic target for the prevention of irreversible fibrosis in degenerative skeletal muscle conditions.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture