MilliporeSigma
  • Theranostic Hyaluronic Acid-Iron Micellar Nanoparticles for Magnetic-Field-Enhanced in vivo Cancer Chemotherapy.
Theranostic Hyaluronic Acid-Iron Micellar Nanoparticles for Magnetic-Field-Enhanced in vivo Cancer Chemotherapy.
ChemMedChem (2017-11-01)
Guohao Wang, Shi Gao, Rui Tian, Jasmine Miller-Kleinhenz, Zainen Qin, Tianji Liu, Lu Li, Fan Zhang, Qingjie Ma, Lei Zhu
ABSTRACT
The delivery of therapeutic cancer agents using nanomaterials has recently attracted much attention. Although encouraging progress with chemotherapeutics has been made, tumor treatment response remains unsatisfactory. To address this concern, we constructed a new micellar nanocomplex by covalently conjugating hyaluronic acid (HA) with an iron oxide nanoparticle (IONP). When an external magnetic field was applied to the tumor area, HA-IONP specifically accumulated in the tumor, due to the strong IONP magnetism. In addition, HA was shown to bind to cluster determinant 44 (CD44), which is overexpressed on tumor cells. With combined magnetic, CD44, and enhanced permeability retention (EPR) targeting, the efficient delivery of HA-IONP to the tumor is expected to enhance cancer treatment efficiency. After encapsulation of the chemotherapy drug homocamptothecin (HCPT), the theranostic potency of HA-IONP/HCPT (HIH) was investigated both in vitro and in vivo. The improved tumor homing behavior of HIH was observed by magnetic resonance imaging (MRI) when an external magnetic field was used. Moreover, HIH showed remarkable tumor ablation efficiency, with magnetic targeting after 3 mg kg