MilliporeSigma
  • Formal total synthesis of oximidine II via a Suzuki-type cross-coupling macrocyclization employing potassium organotrifluoroborates.
Formal total synthesis of oximidine II via a Suzuki-type cross-coupling macrocyclization employing potassium organotrifluoroborates.
Journal of the American Chemical Society (2004-08-19)
Gary A Molander, Florian Dehmel
ABSTRACT
A formal total synthesis of oximidine II has been achieved, employing a Suzuki-type coupling approach to construct the highly strained, polyunsaturated 12-membered macrolactone. To achieve this goal, benefit was derived from the stability of potassium alkenyltrifluoroborates to establish conditions for the macrocyclization. The stereocontrolled formation of the cis-1,2-diol subunit was accomplished using a diastereoselective, reagent controlled addition to a chiral aldehyde utilizing the Carreira protocol. Advantage was taken of the Snieckus hydroborating reagent to gain access to the key trifluoroborate needed for the macrocyclization.