MilliporeSigma
  • Olaratumab in Combination with Doxorubicin for the Treatment of Advanced Soft Tissue Sarcoma: An Evidence Review Group Perspective of a National Institute for Health and Care Excellence Single Technology Appraisal.
Olaratumab in Combination with Doxorubicin for the Treatment of Advanced Soft Tissue Sarcoma: An Evidence Review Group Perspective of a National Institute for Health and Care Excellence Single Technology Appraisal.
PharmacoEconomics (2017-09-16)
Irina A Tikhonova, Tracey Jones-Hughes, James Dunham, Fiona C Warren, Sophie Robinson, Peter Stephens, Martin Hoyle
ABSTRACT
The manufacturer of olaratumab (Lartruvo
Sigma-Aldrich
N,O-Bis(trimethylsilyl)carbamate, ≥98.0% (T)