MilliporeSigma
  • Substrate specificity and subsite affinities of crystalline α-glucosidase from Aspergillus niger
Substrate specificity and subsite affinities of crystalline &#945;-glucosidase from <I>Aspergillus niger</I>
Agricultural and Biological Chemistry (1991)
A. Kita et al