MilliporeSigma
  • Accelerated neointima formation after vascular injury in mice with stromelysin-3 (MMP-11) gene inactivation.
Accelerated neointima formation after vascular injury in mice with stromelysin-3 (MMP-11) gene inactivation.
Arteriosclerosis, thrombosis, and vascular biology (1999-12-11)
H R Lijnen, B Van Hoef, I Vanlinthout, M Verstreken, M C Rio, D Collen
ABSTRACT
The hypothesis that stromelysin-3 (MMP-11), a unique member of the matrix metalloproteinase (MMP) family, plays a role in neointima formation was tested with the use of a vascular injury model in wild-type (MMP-11(+/+)) and MMP-11-deficient (MMP-11(-/-)) mice. Neointima formation 2 to 3 weeks after electric injury of the femoral artery was significantly enhanced in MMP-11(-/-) as compared with MMP-11(+/+) mice, in both mice of a pure 129SV genetic background (0.014 versus 0.0010 mm(2) at 2 weeks, P<0.001) and those of a 50/50 mixed 129SV/BL6 background (0.030 versus 0.013 mm(2) at 3 weeks, P<0.05). The medial areas were comparable, resulting in intima/media ratios that were significantly increased in MMP-11(-/-) as compared with MMP-11(+/+) arteries, in mice of both the 129SV (1. 0 versus 0.18, P<0.001) and mixed (1.5 versus 0.70, P<0.05) backgrounds. Nuclear cell counts in cross-sectional areas of the intima of the injured region were higher in arteries from MMP-11(-/-) mice than in those from MMP-11(+/+) mice (210 versus 48, P<0.001, in pure 129SV mice and 290 versus 150, P<0.01, in mice of the mixed genetic background). Immunocytochemical analysis revealed that alpha-actin-positive and CD45-positive cells were more abundant in intimal sections of MMP-11(-/-) mice. Degradation of the internal elastic lamina was more extensive in arteries of MMP-11(-/-) mice than in those of MMP-11(+/+) mice (39% versus 6.8% at 3 weeks, P<0. 005). The mechanisms by which MMP-11 could impair elastin degradation and cellular migration in this model remain, however, unknown.