MilliporeSigma
  • Prognostic significance of activated Akt expression in pancreatic ductal adenocarcinoma.
Prognostic significance of activated Akt expression in pancreatic ductal adenocarcinoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2004-04-23)
Shinji Yamamoto, Yasuhiko Tomita, Yoshihiko Hoshida, Takaya Morooka, Hiroaki Nagano, Keizo Dono, Koji Umeshita, Masato Sakon, Osamu Ishikawa, Hiroaki Ohigashi, Shoji Nakamori, Morito Monden, Katsuyuki Aozasa
ABSTRACT
Akt is a serine/threonine kinase that plays a central role in tumorigenesis. Among the members of Akt family, Akt2 is associated with the development of human cancers. The present study was designed to clarify the prognostic significance of Akt2 and activated Akt expression in pancreatic ductal adenocarcinoma (PDAC). In addition, activated extracellular signal-regulated kinase 1 and 2 (ERK1/2) and the proliferation activity of tumor cells detected by Ki-67 immunohistochemistry were examined. Immunohistochemical analysis was performed on paraffin-embedded specimens from 65 patients with PDAC; 36 males and 29 females with ages ranging from 48 to 79 years (median, 66 years) of age. Expression levels of Akt2, phosphorylated Akt (p-Akt), and phosphorylated ERK 1/2 (p-ERK 1/2) were categorized as either weaker (low intensity) or equal to stronger (high intensity) compared with those in the endothelial cells of the same specimens. For Ki-67 immunohistochemistry, cases were divided into two groups: level 1, Ki-67 labeling index (LI), <20%; level 2, Ki-67 LI, > or = 20%. Twenty-six (42.6%), 28 (45.9%), 39 (63.9%), and 46 (75.4%) of the tumors showed high intensity of Akt2, p-Akt, and p-ERK 1/2 expression, and Ki-67 LI level 2, respectively. A significant positive correlation was observed between Akt2 and p-Akt expression (P < 0.01). Multivariate analysis revealed that p-Akt expression, Ki-67 LI, and histological differentiation are independent prognosticators for PDAC. p-Akt expression is a significant prognostic indicator for PDAC. Inhibition of Akt is a possible molecular approach for treatment of PDAC.