MilliporeSigma
  • A novel deletion in the GH1 gene including the IVS3 branch site responsible for autosomal dominant isolated growth hormone deficiency.
A novel deletion in the GH1 gene including the IVS3 branch site responsible for autosomal dominant isolated growth hormone deficiency.
The Journal of clinical endocrinology and metabolism (2005-12-22)
Daniela Vivenza, Laura Guazzarotti, Michela Godi, Daniela Frasca, Berardo di Natale, Patricia Momigliano-Richiardi, Gianni Bona, Mara Giordano
ABSTRACT
The majority of mutations responsible for isolated GH type II deficiency (IGHD II) lead to dominant negative deleteriously increased levels of the GH1 exon 3 skipped transcripts. The aim of this study was the characterization of the molecular defect causing a familial case of IGHD II. A 2-yr-old child and her mother with severe growth failure at diagnosis (-5.8 and -6.9 sd score, respectively) and IGHD were investigated for the presence of GH1 mutations. We identified a novel 22-bp deletion in IVS3 (IVS3 del+56-77) removing the putative branch point sequence (BPS). Analysis of patients' lymphocyte mRNA showed an excess exon 3 skipping. The mutated allele transfected into rat pituitary cells produced four differently spliced products: the exon 3 skipped mRNA as the main product and lower amounts of the full-length cDNA and of two novel mRNA aberrant isoforms, one with the first 86 bases of exon 4 deleted and the other lacking the entire exon 4. A mutagenized construct lacking exclusively the 7 bp of the BPS only generated the exon 4 skipped and the full-length isoforms. The presence of the full-length transcript in the absence of the canonical BPS points to an alternative BPS in IVS3. The IVS3 del+56-77 mutation, causing IGHD II in this family, has two separate effects on mRNA processing: 1) exon 3 skipping, analogous to most described cases of IGHD II, an effect likely caused by the reduction in size of the IVS3, and 2) partial or total exon 4 skipping, as a result of the removal of the BPS.