MilliporeSigma
  • Ki-67 staining index predicts distant metastasis and survival in locally advanced prostate cancer treated with radiotherapy: an analysis of patients in radiation therapy oncology group protocol 86-10.
Ki-67 staining index predicts distant metastasis and survival in locally advanced prostate cancer treated with radiotherapy: an analysis of patients in radiation therapy oncology group protocol 86-10.
Clinical cancer research : an official journal of the American Association for Cancer Research (2004-06-26)
Rile Li, Kateri Heydon, M Elizabeth Hammond, David J Grignon, Mack Roach, Harvey B Wolkov, Howard M Sandler, William U Shipley, Alan Pollack
ABSTRACT
Proliferative activity defined by Ki-67 staining index (SI) has been correlated with progression and prognosis in a number of malignant tumors including prostate cancer. However, few studies have examined Ki-67 SI in pretreatment diagnostic material from patients treated with definitive radiotherapy. In a prior study, we found that a Ki-67 SI of >3.5% was associated with poorer patient outcome. The goals of this analysis were to validate the prognostic value of Ki-67 SI and this cut point. Of 456 assessable patients in Radiation Therapy Oncology Group Protocol 86-10, diagnostic material from 108 patients was available for Ki-67 analysis using MIB-1 antibody. Sixty patients were treated with external beam radiotherapy (EBRT) alone, and 48 patients were treated with short-term androgen deprivation + EBRT. Median follow-up was 9 years for those living. The relationship of Ki-67 with distant metastasis (DM), disease-specific survival (DSS), and overall survival (OS) was examined. The median Ki-67 SI was 7.1% (range, 0.2-45.5%). The 7.1% cut point was associated with DM and DSS; however, the 3.5% cut point was as strong a determinant and was the focus of this analysis. In Cox proportional hazards regression, Ki-67 SI was independently associated with DM and DSS. When the Ki-67 SI was </=3.5% and >3.5%, the 5-year risk of DM was 13.5% and 50.8% (P = 0.0005), respectively, and the 5-year risk of DSS was 97.3% and 67.7% (P = 0.0039), respectively. No association of Ki-67 SI with OS was observed. Higher Ki-67 SI was significantly associated with a greater risk of DM and DSS in locally advanced prostate cancer after definitive EBRT or AD + EBRT.