MilliporeSigma
  • HER2/neu testing for anti-HER2-based therapies in patients with unresectable and/or metastatic gastric cancer.
HER2/neu testing for anti-HER2-based therapies in patients with unresectable and/or metastatic gastric cancer.
Journal of clinical pathology (2012-05-10)
Carlos Gómez-Martin, Elena Garralda, M José Echarri, Anabel Ballesteros, Alberto Arcediano, José Luis Rodríguez-Peralto, Manuel Hidalgo, Fernando López-Ríos
ABSTRACT
To study the HER2 gene amplification or overexpression in patients with advanced gastric cancer (GC) and their association with patient characteristics and patient survival. Tumour tissue samples from 148 patients with advanced GC were studied for HER2 by immunohistochemistry (IHC), fluorescence in situ hybridisation (FISH) and dual colour silver enhanced in situ hybridisation (dc-SISH) methods. Clinicopathological data from all patients were collected. Progression free survival and overall survival were also analysed. Mean age was 67 (33-83) years; 75% were male subjects, and 51% had intestinal histological type. HER2+ rates were 10.1% (15/148) by IHC, 18.2% (27/148) by FISH+ or 21.6% (32/148) by dc-SISH+. There were significant differences in HER2+ rates according to histological type when FISH (intestinal, 23%; no intestinal, 4%; p<0.0001) or dc-SISH (intestinal, 26%; no intestinal, 6%; p<0.0001) amplification techniques were used. Median overall survival was significantly longer in HER2+ patients despite the determination technique used: IHC (21.4 vs 9.8 months, HR 0.42; p=0.005); FISH (19.6 vs 9.7 months, HR 0.49; p=0.007) or dc-SISH (19.6 vs 9.7 months, HR 0.53; p=0.009). Factors associated with favourable survival in the multivariate analysis were intestinal type and Her2+ determination by IHC, FISH or dc-SISH. HER2 gene amplification is significantly associated with patient survival. HER2 gene amplification approaches might be an optimal HER2/neu testing strategy for the selection of HER2+ GC patients who are candidates to be treated with anti-HER2 therapies.