MilliporeSigma
HPLC Analysis of Calphostin C on Discovery® HS C18
HPLC Analysis of Calphostin C on Discovery® HS C18 application for HPLC
Materials
related product
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
5 μm particle size, L × I.D. 25 cm × 2.1 mm
CONDITIONS
column
Discovery HS C18, 25 cm × 2.1 mm I.D., 5 μm particles (568503-U)
mobile phase
[A] (0.1% phosphorus acid in water):(0.1% phosphorus acid in acetonitrile); [B] 0.1% phosphorus acid in acetonitrile; (50:50, A:B)
flow rate
0.2 mL/min
column temp.
24 °C
detector
UV, 254 nm
injection
3 μL
sample
Calphostin C (0.25 g/L) in 40% MeOH, 0.025% TFA
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany