MilliporeSigma
HPLC Analysis of Risedronate on SeQuant® ZIC-HILIC
HPLC Analysis of Risedronate on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 100 x 4.6 mm, 5 μm, 200 Å (1.50453.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate
1.0 mL/min
column temp.
ambient
detector
260 nm
injection
50 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany