MilliporeSigma
LC/MS Analysis of Ractopamine on SeQuant® ZIC-HILIC
LC/MS Analysis of Ractopamine on SeQuant® ZIC-HILIC application for LC-MS
CONDITIONS
column
SeQuant® ZIC-cHILIC, PEEK, 150 x 4.6 mm, 3 μm, 100 Å (1.50661.0001)
mobile phase
[A] acetonitrile; [B] 1.361 g of monopotassium phosphate in 1000 ml water (10mM); (80:20, A:B)
flow rate
1.0 mL/min
pressure
913.5 psi 63 Bar
column temp.
35 °C
detector
306 nm cell: 13 μL
injection
5 μL
sample
50 ppb Ractopamine in mobile phase
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany