Skip to Content
MilliporeSigma

AMBH2D6F3F32

Ambeed, Inc.

1-Bromo-2-iodo-4-(trifluoromethyl)benzene

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
350.9
MDL number:

Iupac Name

1-bromo-2-iodo-4-(trifluoromethyl)benzene

Inchi Code

1S/C7H3BrF3I/c8-5-2-1-4(3-6(5)12)7(9,10)11/h1-3H

InChI key

NBSGEAPKXCFNTH-UHFFFAOYSA-N

storage temp.

Keep in dark place, sealed in dry, 2-8C

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Colorless to Yellow-brown Solid or Liquid

Product Link

https://www.ambeed.com/products/640280-28-0.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/640280-28-0.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library