Skip to Content
MilliporeSigma

ENA394767562

Enamine

2-(3-fluorobenzoyl)-1-methyl-1H-imidazole

Synonym(s):

(3-fluorophenyl)(1-methyl-1H-imidazol-2-yl)methanone

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
204.2
MDL number:

Inchi Code

1S/C11H9FN2O/c1-14-6-5-13-11(14)10(15)8-3-2-4-9(12)7-8/h2-7H,1H3

Inchi Key

SZILLIUUSIODKS-UHFFFAOYSA-N

Storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

MSDS

https://www.enaminestore.com/catalog/EN300-128857


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library