Skip to Content
MilliporeSigma

SY3H94E1CB15

Synthonix Corporation

3,4-Difluorobenzylzinc bromide, 0.50 M in THF

Synonym(s):

bromo(3,4-difluorobenzyl)zinc

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
272.41
MDL number:

Inchi Code

1S/C7H5F2.BrH.Zn/c1-5-2-3-6(8)7(9)4-5;;/h2-4H,1H2;1H;/q;;+1/p-1

Inchi Key

UKELZHIDYOTVCK-UHFFFAOYSA-M

Storage temp.

Refrigerator

Purity

0.50

Country of Origin

US

Shipping Temperature

Room Temperature

Physical Form

Solution


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library