Product Name
Tributylmethylammonium bromide ≥98.0%
Product Number
90802
Product Brand
ALDRICH
CAS Number
37026-88-3
Molecular Weight
280.29
Test | Specification |
|---|---|
| APPEARANCE (COLOR) | Colorless or White |
| APPEARANCE (FORM) | Powder or Crystals |
| ARGENTOMETR. TITRATION | 98.0 - 102.0 % |
| SOLUBILITY (COLOR) | Colorless |
| SOLUBILITY (TURBIDITY) | Clear |
| SOLUBILITY (METHOD) | 1G IN 10ML WATER |
| WATER | ≤ 2.0 % |
| CARBON CONTENT | 55.71 % (THEORY) |
| HYDROGEN CONTENT | 10.79 % (THEORY) |
| NITROGEN CONTENT | 5.00 % (THEORY) |