Skip to Content
MilliporeSigma
4-Chloro-7-fluoroquinoline AldrichCPR

4-Chloro-7-fluoroquinoline

4-Chloro-7-fluoroquinoline AldrichCPR
Empirical Formula (Hill Notation):
C9H5ClFN
Molecular Weight:
181.59

Products

Product Category
Brand
Physical Form
Showing 1-2 of 2
Compare
Product Number
Description
Pricing