Skip to Content
MilliporeSigma
2-bromo-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid, 2090987-48-5, undefined,

2-bromo-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid

2-bromo-5,6-dihydro-4H-pyrrolo[1,2-b]pyrazole-3-carboxylic acid, 2090987-48-5, undefined,
CAS Number:
Molecular Weight:
231.05

Products

Brand
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing