Skip to Content
Products
Cart
0
US
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Copper
Linear Formula:
Cu
CAS Number:
7440-50-8
Molecular Weight:
63.55
Products
Shipping
Ships Today
Product Category
metals
Brand
Sigma-Aldrich
Physical Form
Purity
Diameter (mm)
Quality Segment
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
326429
Copper,
wire, diam. 0.25 mm, 99.999% trace metals basis
Expand
View Pricing
An unknown error has occured.