Skip to Content
MilliporeSigma
Methyl orange solution 0.1% indicator pH 3.1-4.4 (red yellow-orange)

Methyl orange solution 0.1%

Methyl orange solution 0.1% indicator pH 3.1-4.4 (red yellow-orange)
Synonym(s):
Methyl orange 0.1% solution

Products

Product Category
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing