Skip to Content
MilliporeSigma
MILLIPLEX® Human Complement Panel

MILLIPLEX® Human Complement Panel

MILLIPLEX® Human Complement Panel
Synonym(s):
Human complement multiplex kit, Luminex human complement immunoassay, Millipore human complement panel, Human membrane attack complex,, human MAC panel, Luminex human complement panel, terminal complement complex, MAC, membrane attack complex

Products

Product Category
Brand
Product Line
Detection Method
Quality Segment
Species Reactivity
Technique
Showing 1-3 of 3