Skip to Content
MilliporeSigma
Normal Mouse IgG Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.

Normal Mouse IgG

Normal Mouse IgG Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Synonym(s):
Mouse IgG Antibody

Products

Product Category
Brand
Biological Source
Clonality
Physical Form
Conjugate
Quality Segment
Technique