Skip to Content
MilliporeSigma
Progesterone solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Progesterone solution

Progesterone solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
EC Number:

Products

Product Category
Brand
Manufacturer
Physical Form
Quality Segment
Application
Special Grade
Technique