MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Design, synthesis and prostate cancer cell-based studies of analogs of the Rho/MKL1 transcriptional pathway inhibitor, CCG-1423.
Design, synthesis and prostate cancer cell-based studies of analogs of the Rho/MKL1 transcriptional pathway inhibitor, CCG-1423.
Bioorganic & medicinal chemistry letters (2009-12-08)
Chris R Evelyn, Jessica L Bell, Jenny G Ryu, Susan M Wade, Andrew Kocab, Nicole L Harzdorf, H D Showalter, Richard R Neubig, Scott D Larsen
ABSTRACT
We recently identified bis(amide) CCG-1423 (1) as a novel inhibitor of RhoA/C-mediated gene transcription that is capable of inhibiting invasion of PC-3 prostate cancer cells in a Matrigel model of metastasis. An initial structure-activity relationship study focusing on bioisosteric replacement of the amides and conformational restriction identified two compounds, 4g and 8, with improved selectivity for inhibition of RhoA/C-mediated gene transcription and attenuated cytotoxicity relative to 1. Both compounds were also capable of inhibiting cell invasion with equal efficacy to 1 but with less attendant cytotoxicity.
Sigma-Aldrich
CCG-100602, ≥98% (HPLC)