MilliporeSigma
  • Poke but don't pinch: risk assessment and venom metering in the western black widow spider, Latrodectus hesperus.
Poke but don't pinch: risk assessment and venom metering in the western black widow spider, Latrodectus hesperus.
Animal behaviour (2014)
Nelsen DR