MilliporeSigma
  • Lipoprotein lipase: biosynthesis, regulatory factors, and its role in atherosclerosis and other diseases
Lipoprotein lipase: biosynthesis, regulatory factors, and its role in atherosclerosis and other diseases
Clinica Chimica Acta; International Journal of Clinical Chemistry (2018)
He PP, et al.