MilliporeSigma
  • Heat Capacity and DC-Magnetic Susceptibility Evidence for the Asymmetry of Electron Spin-Flip Phase Transition of N+ H? O-Bond in Chiral Alanine Crystal
Heat Capacity and DC-Magnetic Susceptibility Evidence for the Asymmetry of Electron Spin-Flip Phase Transition of N+ H? O-Bond in Chiral Alanine Crystal
Acta Physico-Chimica Sinica (2012)
WANG WQ, et al.