MilliporeSigma
  • A midgut lysate of the Riptortus pedestris has antibacterial activity against LPS O-antigen-deficient Burkholderia mutants
A midgut lysate of the Riptortus pedestris has antibacterial activity against LPS O-antigen-deficient Burkholderia mutants
Developmental and Comparative Immunology (2017)
Am Jang H, et al.