MilliporeSigma
  • A practical and chemoselective Mo-catalysed sulfoxide reduction protocol using a 3-mercaptopropyl-functionalized silica gel (MPS)
A practical and chemoselective Mo-catalysed sulfoxide reduction protocol using a 3-mercaptopropyl-functionalized silica gel (MPS)
Royal Society of Chemistry Advances (2016)
Garcia N, et al.