MilliporeSigma
  • Necrostatin-1 analogues: critical issues on the specificity, activity and in vivo use in experimental disease models
Necrostatin-1 analogues: critical issues on the specificity, activity and in vivo use in experimental disease models
Cell Death & Disease (2012)
Takahashi N, et al.