MilliporeSigma
  • New dammarane triterpenoid saponins from the leaves of Cyclocarya paliurus.
New dammarane triterpenoid saponins from the leaves of Cyclocarya paliurus.
Journal of Asian natural products research (2018-04-05)
Yi-Rui Wang, Bao-Song Cui, Shao-Wei Han, Shuai Li
ABSTRACT
Three new dammarane triterpenoid saponins, cyclocariosides O-Q (1-3), were isolated from the ethanolic extracts of the leaves of Cyclocarya paliurus. The structures of these compounds were elucidated by spectroscopic methods.