MilliporeSigma
  • Suzuki-Miyaura cross-coupling reaction of aryl bromides catalyzed by palladium(II) pyridoxal hydrazone complexes
Suzuki-Miyaura cross-coupling reaction of aryl bromides catalyzed by palladium(II) pyridoxal hydrazone complexes
Journal of Organometallic Chemistry (2012)
Pandiarajan, D.; Ramesh, R.