MilliporeSigma
  • Immediate-type allergic and protease-mediated reactions are involved in scratching behaviour induced by topical application of Dermatophagoides farinae extract in NC/Nga mice.
Immediate-type allergic and protease-mediated reactions are involved in scratching behaviour induced by topical application of Dermatophagoides farinae extract in NC/Nga mice.
Experimental dermatology (2017-02-14)
Yoshihito Yamada, Yuhki Ueda, Aki Nakamura, Shoji Kanayama, Rie Tamura, Kei Hashimoto, Tatsumi Matsumoto, Ritsuko Ishii
ABSTRACT
Atopic dermatitis (AD)-like dermatitis can be induced by repeated topical application of an ointment containing Dermatophagoides farinae body (Dfb) extract in NC/Nga mice. This AD-like murine model also exhibits a biphasic increase in the number of scratching behaviour after topical application of Dfb ointment. In this study, we investigated the possible mechanisms underlying the scratching behaviour in each phase. An increase in the content of mast cell-derived mediators such as histamine and 5-hydroxytryptamine in the lesional skin and increased vascular permeability were observed in the early phase after the Dfb ointment application. Chlorpheniramine (H1 receptor antagonist) and cromoglycate (mast cell stabilizer) reduced the scratching behaviour in the early phase but not that in the later phase. Furthermore, the content of various endogenous pruritogens such as interleukin-31 and thymic stromal lymphopoietin in the lesional skin was increased 1 or 24 hours after the Dfb ointment application. Elevated expression of proteinase-activated receptor-2 (PAR-2) was also observed in the epidermis. Finally, gabexate (serine protease inhibitor) reduced the scratching behaviour in both phases, and anti-PAR2 antibody also showed a tendency to reduce both scratching behaviours. These findings suggest that immediate-type allergic reactions caused by mast cell degranulation and PAR-2 activation by proteases are involved in the scratching behaviour in this AD-like model.